ENAH11DD211F Enamine

8-oxa-5-azaspiro[3.6]decane

  • MDL number MFCD31689929
  • Cas Number : 1251713-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1722997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722997
Availability details for
Quantity
  • Availability
  • Add to Cart