ENAH11DD2121 Enamine

3-(aminomethyl)-1-methyl-1H-pyrazole-5-carboxamide hydrochloride

  • MDL number MFCD31689930
  • Cas Number : 232282-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-1722999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722999
Availability details for
Quantity
  • Availability
  • Add to Cart