ENAH11DD2122 Enamine

(2S)-3-fluoro-2-hydroxypropanoic acid

  • MDL number MFCD19230775
  • Cas Number : 97643-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1721269

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721269
Availability details for
Quantity
  • Availability
  • Add to Cart