ENAH11DD212A Enamine

2-tert-butyl-4-methyl-1H-imidazole

  • MDL number MFCD31666713
  • Cas Number : 876310-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1723039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723039
Availability details for
Quantity
  • Availability
  • Add to Cart