ENAH11DD2130 Enamine

2-[(E)-2-nitroethenyl]-1H-imidazole hydrochloride

  • MDL number MFCD31666771
  • Cas Number : 2225181-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-1723050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723050
Availability details for
Quantity
  • Availability
  • Add to Cart