ENAH11DD2132 Enamine

4,4-dimethoxypentan-1-amine

  • MDL number MFCD19207114
  • Cas Number : 142611-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723068

Safety Information

signalword Danger
pcodes P210,P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P370+P378,P403+P235,P405,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-1723068
pictograms GHS05
hstatements H227,H314
Availability details for
Quantity
  • Availability
  • Add to Cart