ENAH11DD2135 Enamine

2-(4-iodo-1H-pyrazol-1-yl)-6-(trifluoromethyl)pyridine

  • MDL number MFCD20569018
  • Cas Number : 1466058-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1723085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723085
Availability details for
Quantity
  • Availability
  • Add to Cart