ENAH11DD2138 Enamine

3-(2,5-dichlorobenzenesulfinyl)propanoic acid

  • MDL number MFCD14613016
  • Cas Number : 1249655-47-7
  • Molecular Weight : 267.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[(2,5-dichlorophenyl)sulfinyl]propanoic acid
Inchi Code 1S/C9H8Cl2O3S/c10-6-1-2-7(11)8(5-6)15(14)4-3-9(12)13/h1-2,5H,3-4H2,(H,12,13)
Inchi Key QBNJIMOKEXOFNS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1721279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721279
Availability details for
Quantity
  • Availability
  • Add to Cart