ENAH11DD2140 Enamine

ethyl 1-(carbamothioylmethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD24643974
  • Cas Number : 1190234-57-1
  • Molecular Weight : 213.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 1-(2-amino-2-thioxoethyl)-1H-pyrazole-4-carboxylate
Inchi Code 1S/C8H11N3O2S/c1-2-13-8(12)6-3-10-11(4-6)5-7(9)14/h3-4H,2,5H2,1H3,(H2,9,14)
Inchi Key DFQLEGDDLVDFPY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1723102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723102
Availability details for
Quantity
  • Availability
  • Add to Cart