ENAH11DD2142 Enamine

1-(4-cyclopropylphenyl)ethan-1-ol

  • MDL number MFCD23806193
  • Cas Number : 40641-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1721284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721284
Availability details for
Quantity
  • Availability
  • Add to Cart