ENAH11DD2143 Enamine

3-methoxypentanoic acid

  • MDL number MFCD17287383
  • Cas Number : 100862-27-9
  • Molecular Weight : 132.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methoxypentanoic acid
Inchi Code 1S/C6H12O3/c1-3-5(9-2)4-6(7)8/h5H,3-4H2,1-2H3,(H,7,8)
Inchi Key ZRWJEWKABOKXBQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1723115

Safety Information

signalword Danger
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1723115
pictograms GHS05,GHS07
hstatements H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart