ENAH11DD2144 Enamine

tert-butyl 5-(chlorosulfonyl)-1,2,3,4-tetrahydroquinoline-1-carboxylate

  • MDL number MFCD28388181
  • Cas Number : 2090008-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1723118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723118
Availability details for
Quantity
  • Availability
  • Add to Cart