ENAH11DD214E Enamine

1-azacycloundecan-2-one

  • MDL number MFCD24503763
  • Molecular Weight : 169.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name azacycloundecan-2-one
Inchi Code 1S/C10H19NO/c12-10-8-6-4-2-1-3-5-7-9-11-10/h1-9H2,(H,11,12)
Inchi Key WULPUQZASCZTPO-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1721295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721295
Availability details for
Quantity
  • Availability
  • Add to Cart