ENAH11DD2151 Enamine

6-chloro-2,4-dimethoxypyridine-3-carbaldehyde

  • MDL number MFCD31666117
  • Cas Number : 2225146-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1720870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720870
Availability details for
Quantity
  • Availability
  • Add to Cart