ENAH11DD2152 Enamine

2-amino-2-(3-chloro-2,4-difluorophenyl)acetic acid hydrochloride

  • MDL number MFCD31666617
  • Cas Number : 2228109-37-1
  • Molecular Weight : 258.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-2-(3-chloro-2,4-difluorophenyl)acetic acid hydrochloride
Inchi Code 1S/C8H6ClF2NO2.ClH/c9-5-4(10)2-1-3(6(5)11)7(12)8(13)14;/h1-2,7H,12H2,(H,13,14);1H
Inchi Key UICQWGFWRALTNB-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1723170

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1723170
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart