ENAH11DD2153 Enamine

3-methyl-2-oxo-1,2,3,4-tetrahydroquinoline-3-carboxylic acid

  • MDL number MFCD24495829
  • Cas Number : 131781-59-4
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methyl-2-oxo-1,2,3,4-tetrahydroquinoline-3-carboxylic acid
Inchi Code 1S/C11H11NO3/c1-11(10(14)15)6-7-4-2-3-5-8(7)12-9(11)13/h2-5H,6H2,1H3,(H,12,13)(H,14,15)
Inchi Key ZSTNDRNEHHDPQB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1720873

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1720873
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart