ENAH11DD2154 Enamine

1-(piperidin-4-yl)-1H-pyrazole-3-carboxylic acid hydrochloride

  • MDL number MFCD31666767
  • Cas Number : 2228472-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-1723191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723191
Availability details for
Quantity
  • Availability
  • Add to Cart