ENAH11DD2157 Enamine

7-bromo-3,4-dihydro-2H-1lambda6,3-benzothiazine-1,1,4-trione

  • MDL number MFCD28140767
  • Cas Number : 1823553-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721315
Availability details for
Quantity
  • Availability
  • Add to Cart