ENAH11DD2158 Enamine

7-bromo-1H,2H,3H,4H-pyrido[2,3-b]pyrazin-3-one

  • MDL number MFCD13180542
  • Cas Number : 957198-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 263-265
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1720876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720876
Availability details for
Quantity
  • Availability
  • Add to Cart