ENAH11DD215B Enamine

1-(azetidin-3-yl)azetidin-3-ol; bis(trifluoroacetic acid)

  • MDL number MFCD31666804
  • Cas Number : 2229570-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2CF3COOH
Product Link https://www.enaminestore.com/catalog/EN300-1723254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723254
Availability details for
Quantity
  • Availability
  • Add to Cart