ENAH11DD215E Enamine

ethyl 2-amino-3-(4-methoxyphenyl)propanoate hydrochloride

  • MDL number MFCD31666615
  • Cas Number : 79751-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-153
Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-1723260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723260
Availability details for
Quantity
  • Availability
  • Add to Cart