ENAH11DD2161 Enamine

1-[(5-methyl-1,3,4-thiadiazol-2-yl)methyl]piperazine dihydrochloride

  • MDL number MFCD31665150
  • Cas Number : 2225147-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-1721320

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721320
Availability details for
Quantity
  • Availability
  • Add to Cart