ENAH11DD2188 Enamine

5-bromo-3-(chloromethyl)-2-fluoropyridine

  • MDL number MFCD15528906
  • Cas Number : 1227584-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720885
Availability details for
Quantity
  • Availability
  • Add to Cart