ENAH11DD218B Enamine

2-azaspiro[4.4]nonane-3-carboxylic acid hydrochloride

  • MDL number MFCD31666690
  • Cas Number : 2228132-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723501
Availability details for
Quantity
  • Availability
  • Add to Cart