ENAH11DD219C Enamine

4-bromo-1-[4-(trifluoromethyl)cyclohexyl]-1H-pyrazol-5-amine

  • MDL number MFCD20108182
  • Cas Number : 1484135-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721422

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P312,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-1721422
pictograms GHS07
hstatements H303,H313,H315,H319,H333,H335
Availability details for
Quantity
  • Availability
  • Add to Cart