ENAH11DD219D Enamine

ethyl 5-bromo-2-methyl-1,3-oxazole-4-carboxylate

  • MDL number MFCD10566060
  • Cas Number : 1260861-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720888
Availability details for
Quantity
  • Availability
  • Add to Cart