ENAH11DD21A4 Enamine

3,3-dimethylhex-5-enoic acid

  • MDL number MFCD19228557
  • Cas Number : 77290-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723674

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P405,P501
hcodes C,Xn
MSDS https://www.enaminestore.com/catalog/EN300-1723674
pictograms GHS05,GHS07
hstatements H302,H314
Availability details for
Quantity
  • Availability
  • Add to Cart