ENAH11DD21A9 Enamine

2-(1H-1,2,3-triazol-1-yl)ethanethioamide

  • MDL number MFCD31666646
  • Cas Number : 2022397-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723685

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723685
Availability details for
Quantity
  • Availability
  • Add to Cart