ENAH11DD21AA Enamine

(3aR,7aS)-octahydro-1H-isoindole-2-sulfonamide

  • MDL number MFCD31666732
  • Cas Number : 26918-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723689
Availability details for
Quantity
  • Availability
  • Add to Cart