ENAH11DD21C8 Enamine

5,6,7,8-tetrahydronaphthalene-2-carbonitrile

  • MDL number MFCD14534426
  • Cas Number : 17104-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 20-21
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723792

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P305+P351+P338,P311,P312,P330,P332+P313,P337+P313,P361,P362,P363,P403+P233,P405,P501
hcodes T,Xi
MSDS https://www.enaminestore.com/catalog/EN300-1723792
pictograms GHS06
hstatements H301,H311,H315,H319,H331,H335
Availability details for
Quantity
  • Availability
  • Add to Cart