ENAH11DD21D6 Enamine

4-(1,1,2-trifluoroethyl)benzoic acid

  • MDL number MFCD31665267
  • Cas Number : 2228364-16-5
  • Molecular Weight : 204.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1723888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723888
Availability details for
Quantity
  • Availability
  • Add to Cart