ENAH11DD21EA Enamine

2,3-bis(trifluoromethyl)cyclopropan-1-amine hydrochloride

  • MDL number MFCD31665269
  • Cas Number : 2228364-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-1723947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723947
Availability details for
Quantity
  • Availability
  • Add to Cart