ENAH11DD21ED Enamine

(4R)-6-fluoro-3,4-dihydrospiro[1-benzopyran-2,1'-cyclopentane]-4-ol

  • MDL number MFCD31666109
  • Cas Number : 939411-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720773
Availability details for
Quantity
  • Availability
  • Add to Cart