ENAH11DD21EE Enamine

7,7-dimethyl-5H,7H-furo[3,4-d]pyrimidin-2-amine

  • MDL number MFCD29484368
  • Cas Number : 1881756-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721491
Availability details for
Quantity
  • Availability
  • Add to Cart