ENAH11DD21EF Enamine

1-(3-aminopropyl)-1,3-diazinan-2-one

  • MDL number MFCD16703069
  • Cas Number : 13173-07-4
  • Molecular Weight : 157.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 48-49
Iupac Name 1-(3-aminopropyl)tetrahydro-2(1H)-pyrimidinone
Inchi Code 1S/C7H15N3O/c8-3-1-5-10-6-2-4-9-7(10)11/h1-6,8H2,(H,9,11)
Inchi Key HQXKCSSNQJUVML-UHFFFAOYSA-N

Description

Melting Point 48-49
Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1723991

Safety Information

signalword Danger
pcodes P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1723991
pictograms GHS05,GHS07
hstatements H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart