ENAH11DD21F0 Enamine

N-ethyl-1H-1,2,4-triazol-5-amine

  • MDL number MFCD19216442
  • Cas Number : 89279-89-0
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-ethyl-1H-1,2,4-triazol-5-amine
Inchi Code 1S/C4H8N4/c1-2-5-4-6-3-7-8-4/h3H,2H2,1H3,(H2,5,6,7,8)
Inchi Key NFSSJGIUWDEDGU-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1723992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723992
Availability details for
Quantity
  • Availability
  • Add to Cart