ENAH11DD21F3 Enamine

5-(acetamidomethyl)-1-propyl-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD21601450
  • Cas Number : 1365961-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1724001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724001
Availability details for
Quantity
  • Availability
  • Add to Cart