ENAH11DD21F4 Enamine

6-ethynyl-3-methoxypyridine-2-carbaldehyde

  • MDL number MFCD22191112
  • Cas Number : 1824166-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1720734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720734
Availability details for
Quantity
  • Availability
  • Add to Cart