ENAH11DD21FB Enamine

N-hydroxy-1H-indazole-3-carboxamide

  • MDL number MFCD16620125
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-hydroxy-1H-indazole-3-carboxamide
Inchi Code 1S/C8H7N3O2/c12-8(11-13)7-5-3-1-2-4-6(5)9-10-7/h1-4,13H,(H,9,10)(H,11,12)
Inchi Key QNYDFCGJGMCOAP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1724019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724019
Availability details for
Quantity
  • Availability
  • Add to Cart