ENAH11DD21FC Enamine

3-{[(tert-butoxy)carbonyl]amino}-5-chlorothiophene-2-carboxylic acid

  • MDL number MFCD31666851
  • Cas Number : 2229142-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1724020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724020
Availability details for
Quantity
  • Availability
  • Add to Cart