ENAH11DD21FD Enamine

6-bromo-4-chloro-1,3-dioxaindane

  • MDL number MFCD23709710
  • Cas Number : 72736-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1721526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721526
Availability details for
Quantity
  • Availability
  • Add to Cart