ENAH11DD2201 Enamine

N-methyl-4-sulfanylbenzamide

  • MDL number MFCD24556421
  • Cas Number : 187160-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-132
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1724076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724076
Availability details for
Quantity
  • Availability
  • Add to Cart