ENAH11DD2204 Enamine

5-methylpiperidine-3-carboxamide hydrochloride

  • MDL number MFCD31665342
  • Cas Number : 2228678-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-1724086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724086
Availability details for
Quantity
  • Availability
  • Add to Cart