ENAH11DD2206 Enamine

N-[2-(1-hydroxyethyl)phenyl]propanamide

  • MDL number MFCD12186329
  • Cas Number : 1156642-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1724088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724088
Availability details for
Quantity
  • Availability
  • Add to Cart