ENAH11DD2209 Enamine

2-methyl-4H,5H,6H,7H,8H-[1,3]thiazolo[5,4-b]azepine hydrochloride

  • MDL number MFCD31666193
  • Cas Number : 2225142-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720910
Availability details for
Quantity
  • Availability
  • Add to Cart