ENAH11DD220B Enamine

5-oxaspiro[3.4]octane-2-carboxylic acid

  • MDL number MFCD31666878
  • Cas Number : 1374658-88-4
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-oxaspiro[3.4]octane-2-carboxylic acid
Inchi Code 1S/C8H12O3/c9-7(10)6-4-8(5-6)2-1-3-11-8/h6H,1-5H2,(H,9,10)
Inchi Key IKQYNEIRTGISNQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1724117

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1724117
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart