ENAH11DD220D Enamine

4-(4-iodophenyl)-1H-pyrazole

  • MDL number MFCD22421405
  • Cas Number : 1399654-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1724120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724120
Availability details for
Quantity
  • Availability
  • Add to Cart