ENAH11DD220F Enamine

5-(difluoromethyl)-2,3-difluoropyridine

  • MDL number MFCD25476751
  • Cas Number : 1804659-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1724122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724122
Availability details for
Quantity
  • Availability
  • Add to Cart