ENAH11DD2212 Enamine

1-methyl-2,3,4,5-tetrahydro-1H-1-benzazepin-5-amine dihydrochloride

  • MDL number MFCD31666194
  • Cas Number : 2225154-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-1720912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720912
Availability details for
Quantity
  • Availability
  • Add to Cart