ENAH11DD2214 Enamine

1-[(3-bromophenyl)methyl]-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD14694479
  • Cas Number : 140480-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1724138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724138
Availability details for
Quantity
  • Availability
  • Add to Cart